missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(1-pyrazolyl)borate, potassium salt, 95%

Product Code. p-198209 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10145560

missing translation for 'mfr': Thermo Scientific Acros 300160010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14782-58-2 | C12H12BKN8 | 318.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14782-58-2
Molecular Formula C12H12BKN8
Molecular Weight (g/mol) 318.2
MDL Number MFCD00800557
InChI Key RKBGLZQVHBLUBM-UHFFFAOYSA-N
Synonym tetrakis 1-pyrazolyl borate, potassium tetrakis pyrazol-1-yloxy boranuide
PubChem CID 92043288
IUPAC Name potassium;tetra(pyrazol-1-yloxy)boranuide
SMILES [B-](ON1C=CC=N1)(ON2C=CC=N2)(ON3C=CC=N3)ON4C=CC=N4.[K+]

Specifications

Melting Point 253°C to 254°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohols,dmf,thf
Formula Weight 318.2
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetrakis(1-pyrazolyl)borate, potassium salt, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.