missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Dried, BHT Stabilized, Reagent Grade, Scharlab

Product Code. 11332575
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11332575 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11332575 Supplier Scharlab Supplier No. TE02232500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains a very low water content of ≤0.005% H2 and stabilized with 250 ppm of 2,6-Di-tert-butyl-4-methylphenol. Tetrahydrofuran, Dried, BHT Stabilized, Reagent Grade, Scharlab is a dry solvent for use in both organic and inorganic synthesis.

Reagent grade solvents are the most frequently used grade for laboratory analytical work. They correspond to a grade of high quality chemicals for laboratory and specialized industrial use.

Specifications

Melting Point -108.5°C
CAS 109-99-9
Color Colorless
Acidity Max. 0.0003meq/g
Alkalinity Max. 0.0002meq/g
Appearance Clear
Density 0.89g/cm3
Quantity 2.5 L
Flash Point −21.5°C
Identification Passes test
Ignition Point 215°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.025%
Molecular Formula C4H8O
Vapor Pressure 173 hPa
MDL Number MFCD00005356
UN Number 2056
Water Max. 0.005%, Dried
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
Boiling Point 65°C to 66°C
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
PubChem CID 8028
ChEBI CHEBI:26911
Formula Weight 72.11g/mol
Percent Purity ≥99.8%
Grade Reagent
Chemical Name or Material Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran, dried (max. 0,005% H2O), reagent grade, stabilized with 250 ppm of 2,6- Di-tert-buty
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.