missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2-furanpropanol, TRC

Product Code. 30418339
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418339 500 mg
30409134 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418339 Supplier Toronto Research Chemicals Supplier No. TRCT293530500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 767-08-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.18
Synonym 1-(2-Tetrahydrofuryl)-3-propanol, 3-(Tetrahydrofuran-2-yl)propan-1-ol, NSC 2804
IUPAC Name 3-(oxolan-2-yl)propan-1-ol
SMILES OCCCC1CCCO1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H14O2/c8-5-1-3-7-4-2-6-9-7/h7-8H,1-6H2
Formula Weight 130.0994
Chemical Name or Material Tetrahydro-2-furanpropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.