missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabenazine, MedChemExpress
Description
Tetrabenazine (Ro 1-9569) is a reversible inhibitor of the vesicular monoamine transporter VMAT2 with the Kd value of 1.34 nM. Tetrabenazine can be used for research on diseases related to hyperactive movement disorders such as Huntington's disease.
Specifications
Specifications
| Chemical Name or Material | Tetrabenazine |
| CAS | 58-46-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H27NO3 |
| Quantity | 100 mg |
| Synonym | Ro 1-9569 |
| Solubility Information | DMSO : 33.33 mg/mL (105.00 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble) |
| SMILES | O=C1[C@@H](CC(C)C)CN2CCC3=CC(OC)=C(OC)C=C3[C@]2([H])C1.[relative stereochemistry] |
| Molecular Weight (g/mol) | 317.42 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?