missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Antibody (BGN/3G8A4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62943B
This item is not returnable.
View return policy
Description
Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA.
Specifications
| Testosterone | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| BSA conjugated testosterone-3,-CMO | |
| 0.1 mL | |
| Cellular Signaling, Developmental Biology | |
| This product recognises testosterone, a steroid hormone and most potent of the naturally occurring androgens. Testosterone promotes the development and maintenance of male secondary sex characteristics and is produced primarily in the testes, but is also produced in small amounts by the adrenal glands, ovaries and placenta. Synthetic derivatives of testosterone are used therapeutically in the treatment of breast cancer and for hormone replacement therapy. NB100-62943 also recognises 5-a-Dihydrotestosterone (1%) and Androstenedione (1%). | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA | |
| BGN/3G8A4 | |
| ELISA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction