Learn More
IKA™ Test Tube Attachment for IKA Vortex Shaker
Shop All IKA Products
Click to view available options
Holds:
12 x 12 mm Reagent Glasses
18 x 10 mm Reagent Glasses
8 x 16 mm Reagent Glasses
8 x 20 mm Reagent Glasses
Product Type:
Test Tube Adapter Attachment
Unit Size:
Each
Description
- Requires VG 3.3 universal attachment
- Facilitates multi-tube vortexing
Specifications
Specifications
| Holds | 8 x 20 mm Reagent Glasses |
| Tube Size | 20 mm |
| Product Type | Test Tube Adapter Attachment |
| Description | Test tube adapter attachment |
| Model | Model VG 3.35 |
| For Use With (Equipment) | VG 3.3 Universal Attachment, Vortexer |
| No. of Positions | 8 |
| Requires | VG 3.3 Universal Attachment |