missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (Ethyl-d5)(isopropyl)carbamate, TRC

Product Code. 30378928
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378928 10 mg 10mg
30370582 25 mg 25mg
30382231 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30378928 Supplier Toronto Research Chemicals Supplier No. TRCE74927210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl (Ethyl-d5)(isopropyl)carbamate
Molecular Formula C10 D5 H16 N O2
Quantity 10 mg
InChI Formula InChI=1S/C10H21NO2/c1-7-11(8(2)3)9(12)13-10(4,5)6/h8H,7H2,1-6H3/i1D3,7D2
SMILES [2H]C([2H])([2H])C([2H])([2H])N(C(C)C)C(=O)OC(C)(C)C
IUPAC Name tert-butyl N-(1,1,2,2,2-pentadeuterioethyl)-N-propan-2-ylcarbamate
Molecular Weight (g/mol) 192.31
Formula Weight 192.189
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.