missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (6-(3-(N-(4-Chlorophenyl)carbamimidoyl)guanidino)hexyl)carbamate, TRC

Product Code. 30345516
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345516 100 mg 100mg
30339060 25 mg 25mg
30335914 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30345516 Supplier Toronto Research Chemicals Supplier No. TRCB692280100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl (6-(3-(N-(4-Chlorophenyl)carbamimidoyl)guanidino)hexyl)carbamate
Analyte or Component Names tert-Butyl (6-(3-(N-(4-Chlorophenyl)carbamimidoyl)guanidino)hexyl)carbamate
Shipping Condition Room Temperature
Molecular Formula C19 H31 Cl N6 O2
InChI Formula InChI=1S/C19H31ClN6O2/c1-19(2,3)28-18(27)24-13-7-5-4-6-12-23-16(21)26-17(22)25-15-10-8-14(20)9-11-15/h8-11H,4-7,12-13H2,1-3H3,(H,24,27)(H5,21,22,23,25,26)
Solution Type Neat
SMILES CC(C)(C)OC(=O)NCCCCCCNC(=N)NC(=N)Nc1ccc(Cl)cc1
IUPAC Name tert-butyl N-[6-[[N-[N-(4-chlorophenyl)carbamimidoyl]carbamimidoyl]amino]hexyl]carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 410.941
Quantity 100 mg
Formula Weight 410.22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.