Learn More
Thermo Scientific Chemicals TergitolTM 15-S-40, 70% solution in water
Product Code. 16308247 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Unit Size:
1 L
2.50 L
250mL
Product Code. 16308247
Brand: Thermo Scientific Chemicals 464260025
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 68131-40-8 | [C2H4O]n C13H27O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68131-40-8
Molecular Formula [C2H4O]n C13H27O
MDL Number MFCD00132411
SMILES CCCCCCCC(CCCCC)OCCO-*
Specifications
Density 1.0720 g/mL
Quantity 2.5 L
Specific Gravity 1.072
Solubility Information Solubility aq. soln.: soluble.
Chemical Name or Material Tergitol* 15-S-40