missing translation for 'onlineSavingsMsg'
Learn More

TEPP-46, MedChemExpress

Product Code. p-200095895
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493577

Brand: Medchem Express HY18657

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TEPP-46 (ML-265) is a potent and selective pyruvate kinase M2 (PKM2) activator with an AC50 of 92 nM, showing little or no effect on PKM1, PKL and PKR.

TEPP-46 (ML-265) is a potent and selective pyruvate kinase M2 (PKM2) activator with an AC50 of 92 nM, showing little or no effect on PKM1, PKL and PKR.

Specifications

Chemical Name or Material TEPP-46
CAS 1221186-53-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H16N4O2S2
Quantity 50 mg
Synonym ML-265
Solubility Information DMSO : 50 mg/mL (134.24 mM; Need ultrasonic)
SMILES O=C1C(N(C)C2=C3SC(S(C)=O)=C2)=C3C=NN1CC4=CC=CC(N)=C4
Molecular Weight (g/mol) 372.46
Formula Weight 372.46
Percent Purity 95.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.