Learn More
Tepilamide fumarate, MedChemExpress
Product Code. 16495206
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495206 100 mg 100mg
16494278 10 mg 10mg
16417007 5 mg 5mg
16400617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16495206 Supplier Medchem Express Supplier No. HY109105
MedChemExpress Tepilamide fumarate (XP-23829) is an oral fumaric acid ester, acts as a prodrug of monomethyl fumarate, and is used in the research of moderate to severe chronic plaque psoriasis.
Tepilamide fumarate (XP-23829) is an oral fumaric acid ester, acts as a prodrug of monomethyl fumarate, and is used in the research of moderate to severe chronic plaque psoriasis.
Specifications
Chemical Name or Material Tepilamide fumarate
CAS 1208229-58-6
Color White
Purity Grade Notes Research
Molecular Formula C11H17NO5
Quantity 100 mg
Synonym XP-23829
Solubility Information DMSO : ≥ 125 mg/mL (513.85 mM) ∣H2O : 50 mg/mL (205.54 mM; Need ultrasonic)
SMILES O=C(OCC(N(CC)CC)=O)/C=C/C(OC)=O
Molecular Weight (g/mol) 243.26
Formula Weight 243.26
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation