missing translation for 'onlineSavingsMsg'
Learn More

Tepilamide fumarate, MedChemExpress

Product Code. 16400617
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16400617 50 mg 50mg
16495206 100 mg 100mg
16494278 10 mg 10mg
16417007 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16400617 Supplier Medchem Express Supplier No. HY109105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tepilamide fumarate (XP-23829) is an oral fumaric acid ester, acts as a prodrug of monomethyl fumarate, and is used in the research of moderate to severe chronic plaque psoriasis.

Tepilamide fumarate (XP-23829) is an oral fumaric acid ester, acts as a prodrug of monomethyl fumarate, and is used in the research of moderate to severe chronic plaque psoriasis.

Specifications

Chemical Name or Material Tepilamide fumarate
CAS 1208229-58-6
Color White
Purity Grade Notes Research
Molecular Formula C11H17NO5
Quantity 50 mg
Synonym XP-23829
Solubility Information DMSO : ≥ 125 mg/mL (513.85 mM) ∣H2O : 50 mg/mL (205.54 mM; Need ultrasonic)
SMILES O=C(OCC(N(CC)CC)=O)/C=C/C(OC)=O
Molecular Weight (g/mol) 243.26
Formula Weight 243.26
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.