missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir Disoproxil Fumarate, TRC

Product Code. 30007200
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007200 10 mg 10mg
30132877 50 mg 50mg
30135185 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007200 Supplier Toronto Research Chemicals Supplier No. TRCT01850510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202138-50-9
Formula Weight 635.184
InChI Formula InChI=1S/C19H30N5O10P.C4H4O4/c1-12(2)33-18(25)28-9-31-35(27,32-10-29-19(26)34-13(3)4)11-30-14(5)6-24-8-23-15-16(20)21-7-22-17(15)24;5-3(6)1-2-4(7)8/h7-8,12-14H,6,9-11H2,1-5H3,(H2,20,21,22);1-2H,(H,5,6)(H,7,8)/b;2-1+/t14-;/m1./s1
IUPAC Name [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acid
Molecular Formula C19 H30 N5 O10 P . C4 H4 O4
Molecular Weight (g/mol) 635.51
Percent Purity >95%
SMILES CC(C)OC(=O)OCOP(=O)(CO[C@H](C)Cn1cnc2c(N)ncnc12)OCOC(=O)OC(C)C.OC(=O)\C=C\C(=O)O
Synonym Tenofovir Disoproxil Fumarate, 5-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]-2,4,6,8-tetraoxa-5-phosphanonanedioic acid 1,9-bis(1-methylethyl) ester 5-oxide (2E)-2-butenedioate (1:1), 5-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]-2,4,6,8-tetraoxa-5-phosphanonanedioic acid bis(1-methylethyl) ester 5-oxide (2E)-2-butenedioate (1:1), (R)-5-[[2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]-2,4,6,8-tetraoxa-5-phosphanonanedioic acid bis(1-methylethyl) ester 5-oxide (E)-2-butenedioate (1:1), GS 4331-05, PMPA Prodrug, Tenfovir disoproxil fumarate, Tenofovir DF, Tenofovir disoproxil fumarate, Virea, Viread, BPPF
Chemical Name or Material Tenofovir Disoproxil Fumarate
Grade HPLC
Show More Show Less

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.