Learn More
Tenofovir alafenamide, MedChemExpress
Product Code. 16492487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16492487
Brand: Medchem Express HY15232
MedChemExpress Tenofovir alafenamide (GS-7340) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.
Tenofovir alafenamide (GS-7340) is an investigational oral prodrug of Tenofovir. Tenofovir is a HIV-1 nucleotide reverse transcriptase inhibitor.
Specifications
Chemical Name or Material Tenofovir alafenamide
CAS 379270-37-8
Color White
Purity Grade Notes Research
Molecular Formula C21H29N6O5P
Quantity 50 mg
Synonym GS-7340
Solubility Information DMSO : ≥ 31 mg/mL (65.06 mM) ∣H2O : 6.67 mg/mL (14.00 mM; Need ultrasonic)
SMILES NC1=NC=NC2=C1N=CN2C[C@@H](C)OC[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O
Molecular Weight (g/mol) 476.47
Formula Weight 476.47
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus