Learn More
Bochem™ Tempered cast iron Tripods
Click to view available options
Diameter (Metric) Outer:
100 mm
140 mm
Unit Size:
Each
Specifications
Specifications
| Material | Tempered Cast Iron |
| Length (Metric) | 210 mm |
| Diameter (Metric) Outer | 140 mm |
| Product Type | Tripods |
| Quantity | 1 Pk. |