Learn More
Eppendorf™ Temperature Verification System, USB, Single Channel
Product Code. 15602559 Shop All Eppendorf Products
Change view
Click to view available options
:
For use with Mastercycler
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15602559 For use with Mastercycler
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15602559 Supplier Eppendorf™ Supplier No. 0056000003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Suitable for use with Mastercycler for PCR assays.
  • Measuring device without temperature sensor
  • Sensor for 96 well and 384 well blocks, sold separately
  • Simple, rapid verification and adjustment
  • Automatic adjustment with Mastercycler software, version 2.10 or higher
  • Can be third-party adjusted
TRUSTED_SUSTAINABILITY
Specifications
Description USB, single-channel
Product Type Temperature Verification System