missing translation for 'onlineSavingsMsg'
Learn More

Temozolomide, MedChemExpress

Product Code. p-200096738
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16473317

Brand: Medchem Express HY17364

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. Temozolomide is also a proautophagic and proapoptotic agent. Temozolomide is effective against tumor cells that are characterized by low levels of O6-alkylguanine DNA alkyltransferase (OGAT) and a functional mismatch repair system. Temozolomide has antitumor and antiangiogenic effects.

Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. Temozolomide is also a proautophagic and proapoptotic agent. Temozolomide is effective against tumor cells that are characterized by low levels of O6-alkylguanine DNA alkyltransferase (OGAT) and a functional mismatch repair system. Temozolomide has antitumor and antiangiogenic effects.

Specifications

Chemical Name or Material Temozolomide
CAS 85622-93-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C6H6N6O2
Quantity 50 mg
Synonym NSC 362856 CCRG 81045 TMZ
Solubility Information DMSO : 20.83 mg/mL (107.29 mM; Need ultrasonic) ∣H2O : 2.86 mg/mL (14.73 mM; Need ultrasonic)
SMILES O=C(C1=C(N2C=N1)N=NN(C)C2=O)N
Molecular Weight (g/mol) 194.15
Formula Weight 194.15
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.