Learn More
Telmisartan Impurity B, TRC
Specifications
Specifications
| Chemical Name or Material | 4'-[(1,7'-Dimethyl-2'-propyl[2,5'-bi-1H-benzimidazol]-1'-yl)methyl]-[1,1'-Biphenyl]-2-carboxylic Acid 1,1-Dimethylethyl Ester |
| CAS | 1026438-56-1 |
| Molecular Formula | C37 H38 N4 O2 |
| Quantity | 50 mg |
| Synonym | Telmisartan Impurity B |
| InChI Formula | InChI=1S/C37H38N4O2/c1-7-13-33-38-31-22-27(35-39-30-18-10-11-19-32(30)40(35)6)20-24(2)34(31)41(33)23-25-14-12-15-26(21-25)28-16-8-9-17-29(28)36(42)43-37(3,4)5/h8-12,14-22H,7,13,23H2,1-6H3 |
| SMILES | CCCc1nc2cc(cc(C)c2n1Cc3cccc(c3)c4ccccc4C(=O)OC(C)(C)C)c5nc6ccccc6n5C |
| IUPAC Name | tert-butyl 2-[3-[[7-methyl-5-(1-methylbenzimidazol-2-yl)-2-propyl-benzimidazol-1-yl]methyl]phenyl]benzoate |
| Molecular Weight (g/mol) | 570.723 |
| Formula Weight | 570.2995 |