Learn More
Telaprevir, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Telaprevir (VX-950) is a highly selective, reversible, and potent peptidomimetic inhibitor of the HCV NS3-4A protease, the steady-state inhibitory constant (Ki) of Telaprevir is 7 nM against a genotype 1 (H strain) NS3 protease domain plus a NS4A cofactor peptide. Telaprevir inhibits SARS-CoV-2 3CLpro activity.
Specifications
Specifications
| Chemical Name or Material | Telaprevir |
| CAS | 402957-28-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C36H53N7O6 |
| Quantity | 10 mg |
| Synonym | VX-950 |
| Solubility Information | DMSO : ≥ 50 mg/mL (73.55 mM) |
| SMILES | O=C(N([C@@H]1C(N[C@H](C(C(NC2CC2)=O)=O)CCC)=O)C[C@@]3(CCC[C@@]31[H])[H])[C@@H](NC([C@@H](NC(C4=NC=CN=C4)=O)C5CCCCC5)=O)C(C)(C)C |
| Molecular Weight (g/mol) | 679.85 |
| Show More |