missing translation for 'onlineSavingsMsg'
Learn More

Scharlab TCBS Agar

Product Code. 11461877
Click to view available options
:
TCBS Agar; Form: Solid
This item is not returnable. View return policy

Product Code. 11461877

Brand: Scharlab 01567500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

TCBS Agar is used for the selective isolation of Vibrio species.

  • APHA/AOAC/BAM/COMPF/FDA/ISO
  • Solid medium for selective isolation of Vibrio parahaemolyticus
  • According to ISO 8914 and 21872-2 standards.

Specifications

Certifications/Compliance ISO 8914 and 21872-2
Form Solid
Media Type TCBS Agar
Product Type Selective Medium
Target Organism Class Gram-negative Bacteria
Formulation Proteose Peptone10.00 g/L, Yeast Extract 5.00 g/L, Sodium Citrate 10.00 g/L, Sodium Thiosulphate 10.00 g/L, Ox Bile 8.00 g/L, Sucrose 20.00 Sodium Chloride 10.00 g/L, Ferric Citrate 1.000 g/L, Thymol Blue 0.04 g/L, Bromthymol Blue 0.04 g/L, Agar 14.00 g/L
For Use With (Application) Selective Isolation of Vibrio Species
Incubation Conditions 37°C ± 1.0°C
Incubation Time 24 ±3 hr.
pH 8.6 ±0.2
Quantity 500 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.