Learn More
HUBER TC100E-F Model Stainless Steel Immersion Cooler
Click to view available options
Model:
TC100E-F
Unit Size:
Each
Description
- Rapid cooling of fluids;
- Ideal for counter-cooling or as a replacement of dry ice
- LED temperature display
- Easy operation with only three keys
- Compact, space-saving construction
- Robust housing with carrying grips
- High quality stainless steel immersion probe
- Temperature controller, stability ±0.5°C
Specifications
Specifications
| Temperature Range | -100°C to +40°C |
| Model | TC100E-F |
| Voltage | 220/240 V |