missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBX10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 18055811
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055811 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18055811 Supplier Invitrogen™ Supplier No. PA5145131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ETYPLPTTSS GWEPRLGSPF PSGPCTSSTG AQAVAEPTGQ GPKNPRVSRV TVQLEMKPLW EEF Antigen sequence identity: Mouse 78%; Rat 49%.

The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TBX10
Gene Accession No. O75333
Gene Alias dancer; Dc; mmTBX7; T-box 10; T-box 10, pseudogene 1; T-box 13; T-box 7; T-box protein 10; T-box protein 13; T-box transcription factor TBX10; T-box-containing transcriptional activator; TBX10; Tbx10-ps1; Tbx13; TBX7
Gene Symbols TBX10
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 16-78. Recombinant protein control fragment (Product #RP-109898).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.