missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | TBK1 |
---|---|
Concentration | 433 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16866125
|
Proteintech
28397-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16856125
|
Proteintech
28397-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The TBK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TBK1. This antibody recognizes human antigen. The TBK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TBK1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
IgG |
433 μg/mL | |
Polyclonal | |
Rabbit | |
29110 | |
Primary |