Learn More
TBK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28397-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16866125
  • 140.00 EUR / 20µL
Estimated Shipment: 07-06-2024
Description
The TBK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TBK1. This antibody recognizes human antigen. The TBK1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TBK1
Polyclonal
Unconjugated
20 μL
29110
IgG
Western Blot, Immunohistochemistry
433 μg/mL