missing translation for 'onlineSavingsMsg'
Learn More

Tazobactam Sodium Salt-15N3, TRC

Product Code. 30003871
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003871 1 mg 1mg
30011856 10 mg 10mg
30004674 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003871 Supplier Toronto Research Chemicals Supplier No. TRCT0101031MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10H11N15N3NaO5S
Molecular Weight (g/mol) 325.25
SMILES [H][C@@]1(CC2=O)N2[C@@H](C(O[Na])=O)[C@@](C[15N]3C=C[15N]=[15N]3)(C)S1(=O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C10H12N4O5S.Na/c1-10(5-13-3-2-11-12-13)8(9(16)17)14-6(15)4-7(14)20(10,18)19;/h2-3,7-8H,4-5H2,1H3,(H,16,17);/q;+1/p-1/t7-,8+,10+;/m1./s1/i11+1,12+1,13+1;
Percent Purity >95%
Chemical Name or Material Tazobactam Sodium Salt-15N3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.