missing translation for 'onlineSavingsMsg'
Learn More

Tavilermide, MedChemExpress

Product Code. 16457428
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457428 10 mg 10mg
16449907 5 mg 5mg
16437406 100 mg 100mg
16423457 50 mg 50mg
16401719 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16457428 Supplier Medchem Express Supplier No. HY17622

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tavilermide is a selective, partial agonist of TrkA, or a nerve growth factor (NGF) mimetic.

Tavilermide is a selective, partial agonist of TrkA, or a nerve growth factor (NGF) mimetic.

Specifications

Chemical Name or Material Tavilermide
CAS 263251-78-1
Color White
Purity Grade Notes Research
Molecular Formula C24H32N6O11
Quantity 10 mg
Synonym MIM-D3
Solubility Information NH4OH : 50 mg/mL (86.13 mM; ultrasonic and adjust pH to 10 with NH4OH) ∣H2O : 2 mg/mL (3.45 mM; Need ultrasonic)
SMILES O=C(O)CC[C@H](N1)C(N[C@@H](CCCCN)C(N[C@H](C(NCC(O)=O)=O)CCOC2=CC=C([N+]([O-])=O)C=C2C1=O)=O)=O
Molecular Weight (g/mol) 580.54
Formula Weight 580.54
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.