missing translation for 'onlineSavingsMsg'
Learn More

Tauroursodeoxycholic Acid Sodium Salt, TRC

Product Code. 30428071
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428071 500 mg 500mg
30426602 1 g 1g
30427172 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428071 Supplier Toronto Research Chemicals Supplier No. TRCT007810500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tauroursodeoxycholic Acid Sodium Salt
CAS 35807-85-3
Molecular Formula C26 H44 N O6 S . Na
Quantity 500 mg
Synonym 2-[[(3α,5β,7β)-3,7-Dihydroxy-24-oxocholan-24-yl]amino]ethanesulfonic acid sodium salt (1:1), Ethanesulfonic acid, 2-[[(3α,5β,7β)-3,7-dihydroxy-24-oxocholan-24-yl]amino]-, monosodium salt (9CI), Tauroursodeoxycholic acid, sodium salt (6CI,7CI), Sodium tauroursodeoxycholate, Sodium tauroursodesoxycholate
InChI Formula InChI=1S/C26H45NO6S.Na/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29;/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1/t16-,17+,18-,19-,20+,21+,22+,24+,25+,26-;/m1./s1
SMILES [Na+].C[C@H](CCC(=O)NCCS(=O)(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
Molecular Weight (g/mol) 521.69
Formula Weight 521.2787
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.