missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAS2R10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16557751
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16557751 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16557751 Supplier Invitrogen™ Supplier No. PA590261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TSLSIFYFLK IANFSNYIFL WLKSRTNMVL PFMIVFLLIS SLLNFAYIAK ILNDYKTKND TVWDLNMYKS EYFIKQILLN LGVIFFFTLS LITCIFLIIS L; Positive Samples: Mouse brain, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.

This gene product belongs to the family of candidate taste receptors that are members of the G-protein-coupled receptor superfamily. These proteins are specifically expressed in the taste receptor cells of the tongue and palate epithelia. They are organized in the genome in clusters and are genetically linked to loci that influence bitter perception in mice and humans. In functional expression studies, they respond to bitter tastants. This gene maps to the taste receptor gene cluster on chromosome 12p13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAS2R10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Tas2r110
Gene Accession No. Q7M712, Q9JKE8
Gene Alias MGC126811; MGC126813; T2R10; TAS2R10; taste 2 receptor member 10; taste receptor family B member 2; Taste receptor type 2 member 10; taste receptor, family B, member 2; taste receptor, type 2, member 10; TRB2
Gene Symbols Tas2r110
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAS2R10 (NP_0764101).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100310875, 387344
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.