Learn More
Tariquidar, MedChemExpress
Product Code. 16464038
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464038 10 mg 10mg
16410397 50 mg 50mg
16425026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16464038 Supplier Medchem Express Supplier No. HY10550
MedChemExpress Tariquidar (XR9576) is a potent and specific inhibitor of P-glycoprotein (P-gp) with the high affinity (Kd=5.1 nM).
Tariquidar (XR9576) is a potent and specific inhibitor of P-glycoprotein (P-gp) with the high affinity (Kd=5.1 nM).
Specifications
Chemical Name or Material Tariquidar
CAS 206873-63-4
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C38H38N4O6
Quantity 10 mg
Synonym XR9576
Solubility Information DMSO : 25 mg/mL (38.66 mM; ultrasonic and adjust pH to 5 with HCl) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC2=CC=CC=C2N=C1)NC3=CC(OC)=C(OC)C=C3C(NC4=CC=C(CCN5CC6=C(C=C(OC)C(OC)=C6)CC5)C=C4)=O
Molecular Weight (g/mol) 646.73
Formula Weight 646.73
Percent Purity 96.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death