missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taprenepag, MedChemExpress
Description
Taprenepag (CP-544326) is a potent and selective prostaglandin EP(2) agonist with IC50s of 10 and 15 nM for human and rat EP2, respectively. Taprenepag shows selectivity for EP2 over other EP receptors (IC50s>3200 nM for EP1, EP3, and EP4) and a panel of 37 G protein-coupled receptors.
Specifications
Specifications
| Chemical Name or Material | Taprenepag |
| CAS | 752187-80-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H22N4O5S |
| Quantity | 10 mg |
| Synonym | CP-544326 |
| Solubility Information | DMSO : ≥ 100 mg/mL (208.98 mM) |
| SMILES | O=S(C1=CN=CC=C1)(N(CC2=CC(OCC(O)=O)=CC=C2)CC3=CC=C(N4C=CC=N4)C=C3)=O |
| Molecular Weight (g/mol) | 478.52 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?