Learn More
Sonics Materials™ Tapered Microtip
Click to view available options
Diameter (Metric):
5 mm
6 mm
Unit Size:
Each
Specifications
Specifications
| Amplitude | 212 μm |
| Product Type | Basic Disperser |
| Diameter (Metric) | 5 mm |
| Packaging Quantity | 1 Ea. |
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion