missing translation for 'onlineSavingsMsg'
Learn More

Tandospirone, MedChemExpress

Product Code. p-200098828
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16446098

Brand: Medchem Express HY14558

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tandospirone (SM-3997) is a potent and selective 5-HT1A receptor partial agonist, with a Ki of 27 nM. Tandospirone has anxiolytic and antidepressant activities. Tandospirone can be used for the research of the central nervous system disorders and the underlying mechanisms.

Tandospirone (SM-3997) is a potent and selective 5-HT1A receptor partial agonist, with a Ki of 27 nM. Tandospirone has anxiolytic and antidepressant activities. Tandospirone can be used for the research of the central nervous system disorders and the underlying mechanisms.

Specifications

Chemical Name or Material Tandospirone
CAS 87760-53-0
Color White
Purity Grade Notes Research
Molecular Formula C21H29N5O2
Quantity 10 mg
Synonym SM-3997
Solubility Information DMSO : 16.67 mg/mL (43.47 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.26 mM; Need ultrasonic)
SMILES O=C1[C@]2([H])[C@]([C@H]3CC[C@@H]2C3)([H])C(N1CCCCN(CC4)CCN4C5=NC=CC=N5)=O
Molecular Weight (g/mol) 383.49
Formula Weight 383.49
Percent Purity 96.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.