missing translation for 'onlineSavingsMsg'
Learn More

TAN Solvent, Chem-Lab

Product Code. 13427846
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13427846 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13427846 Supplier Chem-Lab Supplier No. CL02.2009.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to determine the sub of all acid compounds present in a sample. TAN Solvent, Chem-Lab (total acid number) contains 50% C7H8 + 49.5% IPA + 0.5% H2O by volume.

For laboratory use according to ASTM D-664, D-974, D-3242, D-3339 for determining Total Acid Number. It is available in a 5L bottle.

Specifications

CAS 67-63-0, 7732-18-5
Ignition Point 425°C
Density 0.83g/mL
Physical Form Liquid
Molecular Formula C7H8
MDL Number MFCD00008512
Color Colorless
Solubility Soluble in toluene, Poorly soluble in water
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
Quantity 5 L
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 5L SOLUTION MEASUREMENT TAN
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: toluene, propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.