Learn More
BRAND™ Tamper-Evident Screw Cap
Shop All Brand Products
Click to view available options
Color:
Blue
Green
Purple
Red
Transparent
Yellow
Unit Size:
Pack of 1000
Description
- With silicone seal
- For microtubes
- Working temperature between -196 to 121°C
Specifications
Specifications
| Includes | Silicone Seal |
| Liner Material | Silicone |
| Product Type | Tamper-evident Screw Cap |
| Material | Polypropylene |
| Color | Transparent |
| For Use With (Equipment) | 78076064 Microtubes |