missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Taltobulin trifluoroacetate
Specifications
Specifications
| Purity | 99.96% |
| CAS | 228266-41-9 |
| Content And Storage | 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 587.67 g/mol |
| Product Type | Reference compound |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : ≥ 39 mg/mL (66.36 mM) |
| Synonym | HTI-286 trifluoroacetate∣ SPA-110 trifluoroacetate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?