missing translation for 'onlineSavingsMsg'
Learn More

Taltirelin, MedChemExpress

Product Code. p-200100207
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494407

Brand: Medchem Express HYB0596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Taltirelin (TA0910) is a superagonist at thyrotropin-releasing hormone receptor (TRH-R) with an IC50 of 910 nM and EC50 of 36 nM for stimulating an increase in cytosolic Ca2+ concentration (Ca2+ release).

Taltirelin (TA0910) is a superagonist at thyrotropin-releasing hormone receptor (TRH-R) with an IC50 of 910 nM and EC50 of 36 nM for stimulating an increase in cytosolic Ca2+ concentration (Ca2+ release).

Specifications

Chemical Name or Material Taltirelin
CAS 103300-74-9
Color White
Purity Grade Notes Research
Molecular Formula C17H23N7O5
Quantity 50 mg
Synonym TA-0910
Solubility Information DMSO : 100 mg/mL (246.66 mM; Need ultrasonic) ∣H2O : 100 mg/mL (246.66 mM; Need ultrasonic)
SMILES O=C(N(CCC1)[C@@H]1C(N)=O)[C@@H](NC([C@@H](NC2=O)CC(N2C)=O)=O)CC3=CN=CN3
Molecular Weight (g/mol) 405.41
Formula Weight 405.41
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.