Learn More
Köttermann Tall Acid and Base Cabinet
Click to view available options
Configuration:
2 Left-hinged Doors, 4 Drawers
2 Right-hinged Doors, 4 Drawers
Includes:
No Ventilation Fan
Ventilation Fan
Unit Size:
Each
Description
- Safe storage of corrosive non-flammable hazardous substances, with acids and bases separated
- Cabinet carcasses and pull-out trays made of resistant polypropylene
- Liquid-tight drawers on low-friction PTFE strips with pull-out stop
- Removal of aggressive vapors via optional ventilation with an external fan
- Corrosion-resistant locks and hinges
- Ideal for installation under high-performance fume cupboards
Specifications
Specifications
| Configuration | 2 Left-hinged Doors, 4 Drawers |
| Dimensions (D x W x H) | 515 x 600 x 1900 mm |
| Door Style | Hinged, Drawer |
| For Use With (Application) | Acid and Base Storage |
| Includes | No Ventilation Fan |
| Color | White |
| Material | Polypropylene |