missing translation for 'onlineSavingsMsg'
Learn More

TAK-243, MedChemExpress

Product Code. 16465777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465777 5 mg 5mg
16493038 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465777 Supplier Medchem Express Supplier No. HY100487

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAK-243 (MLN7243) is a first-in-class, selective ubiquitin activating enzyme, UAE (UBA1) inhibitor (IC50=1 nM), which blocks ubiquitin conjugation, disrupting monoubiquitin signaling as well as global protein ubiquitination. TAK-243 (MLN7243) induces endoplasmic reticulum (ER) stress, abrogates NF-κB pathway activation and promotes apoptosis.

TAK-243 (MLN7243) is a first-in-class, selective ubiquitin activating enzyme, UAE (UBA1) inhibitor (IC50=1 nM), which blocks ubiquitin conjugation, disrupting monoubiquitin signaling as well as global protein ubiquitination. TAK-243 (MLN7243) induces endoplasmic reticulum (ER) stress, abrogates NF-κB pathway activation and promotes apoptosis.

Specifications

Chemical Name or Material TAK-243
Physical Form Solid
CAS 1450833-55-2
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H20F3N5O5S2
For Use With (Application) Cancer-programmed cell death
Synonym MLN7243
Solubility Information DMSO : 50 mg/mL (96.24 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.92 mM; Need ultrasonic)
SMILES O=S(OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](NC2=CC=NC3=CC(C4=CC=CC(SC(F)(F)F)=C4)=NN23)C1)(N)=O
Molecular Weight (g/mol) 519.52
Formula Weight 519.52
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.