missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16619024
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16619024 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16619024 Supplier Invitrogen™ Supplier No. PA5102551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAF9.

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds to the basal transcription factor GTF2B as well as to several transcriptional activators such as p53 and VP16. A similar but distinct gene (TAF9L) has been found on the X chromosome and a pseudogene has been identified on chromosome 19. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAF9
Gene Accession No. Q16594, Q5BKE0, Q8VI33
Gene Alias 2310012M09Rik; AA673500; AD-004; AU045423; CGI-137; CINAP; CIP; EMBL:AAH91109.1}; hCINAP; MGC:1603; MGC:3647; MGC:5067; RNA polymerase II TBP-associated factor subunit G; STAF31/32; TAF2G; TAF9; taf9 {ECO:0000312; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32 kDa; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32kDa; TAF9 RNA polymerase II, TATA box binding protein-associated factor; Tafii31; TAFII-31; TAFII32; TAFII-32; TAFIID32; TATA box binding protein associated factor 9; TATA-box binding protein associated factor 9; transcription initiation factor TFIID 31 kD subunit; transcription initiation factor TFIID 31 kDa subunit; Transcription initiation factor TFIID 32 kDa subunit; transcription initiation factor TFIID subunit 9
Gene Symbols TAF9
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAF9(Accession Q16594), corresponding to amino acid residues K134-G184.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108143, 373541, 6880
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.