Learn More
TAE 684, TRC
Specifications
Specifications
| Chemical Name or Material | TAE 684 |
| CAS | 761439-42-3 |
| Molecular Formula | C30 H40 Cl N7 O3 S |
| Quantity | 5 mg |
| Synonym | 5-Chloro-N2-[2-methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]-N4-[2-[(1-methylethyl)sulfonyl]phenyl]-2,4-pyrimidinediamine, AE 684, NVP-TAE 684, NVT TAE-684, TAE 684 |
| InChI Formula | InChI=1S/C30H40ClN7O3S/c1-21(2)42(39,40)28-8-6-5-7-26(28)33-29-24(31)20-32-30(35-29)34-25-10-9-23(19-27(25)41-4)37-13-11-22(12-14-37)38-17-15-36(3)16-18-38/h5-10,19-22H,11-18H2,1-4H3,(H2,32,33,34,35) |
| SMILES | COc1cc(ccc1Nc2ncc(Cl)c(Nc3ccccc3S(=O)(=O)C(C)C)n2)N4CCC(CC4)N5CCN(C)CC5 |
| IUPAC Name | 5-chloro-2-N-[2-methoxy-4-[4-(4-methylpiperazin-1-yl)piperidin-1-yl]phenyl]-4-N-(2-propan-2-ylsulfonylphenyl)pyrimidine-2,4-diamine |
| Molecular Weight (g/mol) | 614.2 |
| Formula Weight | 613.2602 |