Learn More
TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Product Code. p-4562588 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Purified
Formulation:
Purified w/o BSA and Azide
Purified with BSA and Azide
Unit Size:
100µL
1mL
500µL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form Formulation unitSize
12643107 500 μL Concentrated Purified with BSA and Azide 500µL
12623107 1 mL Concentrated Purified with BSA and Azide 1mL
12663107 1 mL Concentrated Purified w/o BSA and Azide 1mL
12633107 100 μL Purified Purified with BSA and Azide 100µL
12653107 500 μL Purified Purified w/o BSA and Azide 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12643107 Supplier Epredia Supplier No. RB1660P1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provide accurate, reproducible results Epredia™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.
TACE (TNF-α converting enzyme) protein is a membrane bound matalloprotease from ADAM (a disintegrin and metalloprotease) family of proteins. It cleaves 26kDa type II membrane-bound precursor of TNF-α into 17kDA mature form which is secreted. TACE is also known to process other membrane proteins including TNF receptor, TGFα, L-selectin and amyloid protein precursor.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminus of human TACE protein
Molecular Weight: 80-130kDa (corresponding to mature, precursor and glycosylated forms)
Positive Control: HeLa cells
Cellular Localization: Membrane
Recommended for:
  • Western Blotting
  • Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
TRUSTED_SUSTAINABILITY
Specifications
Antigen TACE (TNF-alpha converting enzyme)/ADAM17 Ab-1
Applications Western Blot
Classification Polyclonal
Clone Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified with BSA and Azide
Host Species Rabbit
Immunogen A synthetic peptide from the C-terminus of human TACE protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG