Learn More
TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminus of human TACE protein
Molecular Weight: 80-130kDa (corresponding to mature, precursor and glycosylated forms)
Positive Control: HeLa cells
Cellular Localization: Membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | TACE (TNF-alpha converting enzyme)/ADAM17 Ab-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified with BSA and Azide |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from the C-terminus of human TACE protein |
| Quantity | 500 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.