Learn More
BRAND™ T-Bore Stopcock
Product Code. 10203781 Shop All Brand Products
Click to view available options
Diameter (Metric):
3 mm
4 mm
Unit Size:
Each
Product Code. 10203781
Brand: BRAND™ 88660
Only null left
Add to basket
Add to basket
T-Bore Stopcock is constructed of PTFE and includes a nozzle.
  • Polypropylene valve lever
  • Outstanding chemical resistance
  • With integrated hose nozzles
TRUSTED_SUSTAINABILITY
Specifications
Type T-Bore Stopcock
Diameter (Metric) 3 mm
Material PTFE
For Use With (Equipment) 5 mm inner diameter tubes
Includes Nozzle