missing translation for 'onlineSavingsMsg'
Learn More

SYT11 Monoclonal Antibody (OTI3F3), TrueMAB™, OriGene

Product Code. 17504564
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17504564 30 μL 30µL
17584554 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17504564 Supplier Origene Technologies Supplier No. TA810985S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SYT11 Monoclonal antibody specifically detects SYT11 in Human samples. It is validated for Western Blot

Specifications

Antigen SYT11
Applications Western Blot
Classification Monoclonal
Clone OTI3F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene AZIN1
Gene Accession No. O14977
Gene Alias 1500004A13Rik; 3632445O20Rik; 5430404N14Rik; 6530420C11Rik; AI851970; KIAA0080; synaptotagmin 11; synaptotagmin 12; synaptotagmin XI; Synaptotagmin-11; SYT11; SYT12; SytXI
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 260-448 of human AZIN1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51582
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.