Learn More
CELLPATH SYSTEM II+ BIOPSY CASSETTE
Product Code. p-200113603
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 2000
Pack of 3000
Product Code. 30226243
Brand: CELLPATH EAM060203A
Incorporating 0.9 x 0.9 mm apertures, the System II+ Biopsy Cassette offers improved specimen security for small biopsy and tissue fragments. CellPath's System II+ Cassettes come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Loose
Product Type Cassette