Learn More
Syringin, TRC
Product Code. 30411890
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411890 25 mg -
30411431 5 mg 5mg
30410061 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30411890 Supplier Toronto Research Chemicals Supplier No. TRCS92005025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118-34-3
Molecular Formula C17 H24 O9
Molecular Weight (g/mol) 372.37
Synonym 4-[(1E)-3-Hydroxy-1-propen-1-yl]-2,6-dimethoxyphenyl-β-D-glucopyranoside, Eleutheroside B, Ilexanthin A, Ligustrin, Lilacin, Magnolenin, Methoxyconiferine, NSC 287441, Sinapyl alcohol 4-O-glucoside, Siringin, Syringoside
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[4-[(E)-3-hydroxyprop-1-enyl]-2,6-dimethoxyphenoxy]oxane-3,4,5-triol
SMILES COc1cc(\C=C\CO)cc(OC)c1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C17H24O9/c1-23-10-6-9(4-3-5-18)7-11(24-2)16(10)26-17-15(22)14(21)13(20)12(8-19)25-17/h3-4,6-7,12-15,17-22H,5,8H2,1-2H3/b4-3+/t12-,13-,14+,15-,17+/m1/s1
Formula Weight 372.142
Percent Purity >95%
Chemical Name or Material Eleutheroside B
Grade HPLC