Learn More
Thermo Scientific™ Syringes for Thermo Scientific™ HPLC Instruments
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
Specifications
| Product Type | HPLC Autosampler Syringe |
| Disposable | Reusable |
| Volume (Metric) | 500 μL |
| For Use With (Equipment) | AS1000, AS3000 Autosampler |