Learn More
Poulten & Graf GmbH FORTUNA™ Gas Syringe
Product Code. 10272141
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10272141 50 mL Each
10519841 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10272141 Supplier Poulten & Graf GmbH Supplier No. 2 00046
Attain precision with Volac Smart™ Micropipette. Poulten Graf™ Volac Smart™ Variable Volume Micropipette are perfect for pipetting small amount of liquid samples in the laboratory.
  • Made of borosilicate glass
  • Amber graduations
Specifications
Autoclavable Not Autoclavable
Diameter (Metric) Inner Needle 2.5 mm
Diameter (Metric) Outer Needle 7.5 mm
Product Type Syringe
Quantity 1/Pk.
Volume (Metric) 50 mL