missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYNE4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542662
This item is not returnable.
View return policy
Description
Peptide sequence: GEESTSPEQA QTLGQDSLGP PEHFQGGPRG NEPAAHPPRW STPSSYEDPA Sequence homology: Cow: 85%; Horse: 85%; Human: 100%.
The exact function of C19orf46 remains unknown.
Specifications
| SYNE4 | |
| Polyclonal | |
| Unconjugated | |
| SYNE4 | |
| C18H19orf46; C19orf46; C1H19orf46; deafness, autosomal recessive 76; DFNB76; KASH domain-containing protein 4; KASH domain-containing protein C19orf46 homolog; KASH4; Nesp4; Nesprin-4; nuclear envelope spectrin repeat protein 4; RGD1304580; spectrin repeat containing nuclear envelope family member 4; spectrin repeat containing, nuclear envelope family member 4; Syne4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 163183 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N205 | |
| SYNE4 | |
| Synthetic peptide directed towards the N-terminal of human C19orf46 (aa 38-87). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction