Learn More
SYCP3 Monoclonal Antibody (OTI2F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806615S
This item is not returnable.
View return policy
Description
SYCP3 Monoclonal antibody specifically detects SYCP3 in Human samples. It is validated for Western BlotSpecifications
| SYCP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IZU3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 50511 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2F4 | |
| Unconjugated | |
| Sycp3 | |
| Cor1; LOC615896 protein; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; Sycp3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like | |
| Full length human recombinant protein of human SYCP3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |