Learn More
SYCP3 Monoclonal Antibody (OTI2F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA806615S
Encompass_Preferred
 View more versions of this product
Product Code. 17546863
  • 242.00 EUR / 30µL
Description
SYCP3 Monoclonal antibody specifically detects SYCP3 in Human samples. It is validated for Western Blot
Specifications
Specifications
SYCP3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8IZU3
Mouse
Affinity chromatography
RUO
50511
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2F4
Unconjugated
Sycp3
Cor1; LOC615896 protein; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; Sycp3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like
Full length human recombinant protein of human SYCP3 produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions